6-Chloropurine ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

C0278-25G C0278-5G
TCC0278-25GEA 255.96 CAD
TCC0278-25G TCC0278-005G
6-Chloropurine ≥98.0% (by titrimetric analysis)
6-Chloropurine

CAS Number: 87-42-3
MDL Number: MFCD00075825
Molecular Formula: C5H3ClN4
Molecular Weight: 154.56
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₅H₃ClN₄
Boiling Pt: 185…192 °C (2 mmHg)
Melting Pt: 178…179 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075825
CAS Number: 87-42-3
EINECS: 201-745-6
Merck Index: 13,02180
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f8804410e476380d3c1fccdafc63a467998390b&vid=5f8804410e476380d3c1fccdafc63a467998390b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR