4-Chloro-o-phenylenediamine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

C0255-25G C0255-5G
TCC0255-025GEA 97.96 CAD
TCC0255-025G TCC0255-5G
4-Chloro-o-phenylenediamine ≥97.0% (by GC, titration analysis)
4-Chloro-o-phenylenediamine

CAS Number: 95-83-0
MDL Number: MFCD00011691
Molecular Formula: C6H7ClN2
Molecular Weight: 142.59
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 75


Other Product Names / Synonyms: 3,4-Diaminochlorobenzene

Formula: C₆H₇ClN₂
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 75 °C
MDL Number: MFCD00011691
CAS Number: 95-83-0
EINECS: 202-456-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91c36128791397ab0a188d874a79bc4b8ef7bebd&vid=91c36128791397ab0a188d874a79bc4b8ef7bebd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR