(4-Chlorophenoxy)acetic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

C0250-25G C0250-500G
TCC0250-025GEA 51.44 CAD
TCC0250-025G TCC0250-500G
(4-Chlorophenoxy)acetic acid ≥98.0% (by HPLC, titration analysis)
(4-Chlorophenoxy)acetic acid

CAS Number: 122-88-3
MDL Number: MFCD00004305
Molecular Formula: C8H7ClO3
Molecular Weight: 186.59
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 158


Product Categories: Bioscience, Auxins
Other Product Names / Synonyms: 4-CPA

Formula: (C₆H₄Cl)OCH₂COOH
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 158 °C
Density: 1.53 g/cm³ (20 °C)
MDL Number: MFCD00004305
CAS Number: 122-88-3
EINECS: 204-581-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c0f52ffcf3a490e71a05e385028422be1b64d89&vid=0c0f52ffcf3a490e71a05e385028422be1b64d89&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR