2-Chlorophenothiazine ≥98.0%

Supplier: TCI America

C0248-500G C0248-25G
TCC0248-500GEA 510.81 CAD
TCC0248-500G TCC0248-025G
2-Chlorophenothiazine ≥98.0%
2-Chlorophenothiazine

CAS Number: 92-39-7
MDL Number: MFCD00005016
Molecular Formula: C12H8ClNS
Molecular Weight: 233.71
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 198

Formula: C₁₂H₈ClNS
Melting Pt: 198 °C
MDL Number: MFCD00005016
CAS Number: 92-39-7
EINECS: 202-152-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d71b7bdea08df854e034c40675abb96f1d84e912&vid=d71b7bdea08df854e034c40675abb96f1d84e912&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR