Chlorophenol red ≥93.0%

Supplier: TCI America

Synonyms: 3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide

C0245-5G C0245-1G
TCC0245-5GEA 44.24 CAD
TCC0245-5G TCC0245-001G
Chlorophenol red ≥93.0%
Chlorophenol red

CAS Number: 4430-20-0
MDL Number: MFCD00005877
Molecular Formula: C19H12Cl2O5S
Molecular Weight: 423.26
Purity/Analysis Method: >93.0% (HPLC)
Form: Crystal

Formula: C₁₉H₁₂Cl₂O₅S
MW: 423.27 g/mol
Melting Pt: 261
Storage Temperature: Ambient
MDL Number: MFCD00005877
CAS Number: 4430-20-0
EINECS: 224-619-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9b190b4b4f823ca678a2bd7ef504f8e35116e59&vid=c9b190b4b4f823ca678a2bd7ef504f8e35116e59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 93.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR