(2-Chloroethoxy)benzene ≥95.0%

Supplier: TCI America

Synonyms: ω-Chlorophenetole, 2-Chloroethyl phenyl ether, β-Chlorophenetole, 2-Phenoxyethyl chloride

C0240-25ML C0240-500ML
TCC0240-025MLEA 132.72 CAD
TCC0240-025ML TCC0240-500ML
(2-Chloroethoxy)benzene ≥95.0%
(2-Chloroethoxy)benzene

CAS Number: 622-86-6
MDL Number: MFCD00018946
Molecular Formula: C8H9ClO
Molecular Weight: 156.61
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 220
Flash Point (°C): 107
Freezing point (°C): 24


Other Product Names / Synonyms: 2-Chloroethyl Phenyl Ether, 2-Phenoxyethyl Chloride

Formula: C₆H₅OCH₂CH₂Cl
MW: 156.61 g/mol
Boiling Pt: 220 °C
Flash Pt: 107 °C
MDL Number: MFCD00018946
CAS Number: 622-86-6
EINECS: 210-757-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be79065e42e801268c3bf9e4b9ae8eece6a9b6a2&vid=be79065e42e801268c3bf9e4b9ae8eece6a9b6a2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR