1-Chlorononane ≥98.0%

Supplier: TCI America

C0233-500ML C0233-25ML
TCC0233-500MLEA 505.6 CAD
TCC0233-500ML TCC0233-025ML
1-Chlorononane ≥98.0%
1-Chlorononane

CAS Number: 2473-01-0
MDL Number: MFCD00001023
Molecular Formula: C9H19Cl
Molecular Weight: 162.70
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 204
Flash Point (°C): 74
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Nonyl Chloride

Formula: C₉H₁₉Cl
MW: 162.7 g/mol
Boiling Pt: 203
Melting Pt: -39.4
Density: 0.87 (20 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00001023
CAS Number: 2473-01-0
EINECS: 219-595-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48cd720eb53f004720212c25d875fb99fce2268c&vid=d828c0d85e43425a387b6d7809265448151ca28e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR