5-Chloro-2-nitrobenzoic acid ≥98.0%
Supplier: TCI America
|
Warning
|
C0224-25G
C0224-250G
TCC0224-025GEA
104.28
CAD
TCC0224-025G
TCC0224-250G
5-Chloro-2-nitrobenzoic acid ≥98.0%
5-Chloro-2-nitrobenzoic acid
CAS Number: 2516-95-2
MDL Number: MFCD00007290
Molecular Formula: C7H4ClNO4
Molecular Weight: 201.56
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 139
Flash Point (°C): 100
Formula:
C₇H₄ClNO₄ Melting Pt: 139 °C Density: 1.593 g/cm³ (18 °C) Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007290 CAS Number: 2516-95-2 EINECS: 219-738-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...