4-Chloro-3-nitrobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C0223-500G C0223-25G
TCC0223-500GEA 139.04 CAD
TCC0223-500G TCC0223-25G
4-Chloro-3-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
4-Chloro-3-nitrobenzoic acid

CAS Number: 96-99-1
MDL Number: MFCD00007079
Molecular Formula: C7H4ClNO4
Molecular Weight: 201.56
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 182

Formula: C₇H₄ClNO₄
Melting Pt: 182 °C
Density: 1.645 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007079
CAS Number: 96-99-1
EINECS: 202-550-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e672f341d94b9e47003cac5df93bc436b3804e38&vid=f24c978d7c22ba4508fcf147da8597d5da39dd3a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR