2-Chloro-5-nitrobenzaldehyde ≥96.0% (by titrimetric analysis)

Supplier: TCI America

C0219-25G C0219-250G
TCC0219-025GEA 41.08 CAD
TCC0219-025G TCC0219-250G
2-Chloro-5-nitrobenzaldehyde ≥96.0% (by titrimetric analysis)
2-Chloro-5-nitrobenzaldehyde

CAS Number: 6361-21-3
MDL Number: MFCD00007293
Molecular Formula: C7H4ClNO3
Molecular Weight: 185.56
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Melting point (°C): 80

Formula: C₇H₄ClNO₃
Melting Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00007293
CAS Number: 6361-21-3
EINECS: 228-830-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b97823bddd00085217ae140ef83655b0cb900fdc&vid=b1f7650fe4dc2d48cc1b7bd357fd5d8a0e8aa8c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR