ε-Caprolactam ≥99.0%
Supplier: TCI America
|
Warning
|
Synonyms:
Azepan-2-one, 2-Oxohexamethyleneimine, 6-Aminocaproic acid lactam
C0203-25G
C0203-500G
TCC0203-025GEA
31.71
CAD
TCC0203-025G
TCC0203-500G
ε-Caprolactam ≥99.0%
ε-Caprolactam
CAS Number: 105-60-2
MDL Number: MFCD00006936
Molecular Formula: C6H11NO
Molecular Weight: 113.16
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 267
Melting point (°C): 70
Flash Point (°C): 130
Other Product Names / Synonyms: Cyclohexanoneisooxime, 2-Oxohexamethyleneimine
Formula:
C₆H₁₁NO Boiling Pt: 267 °C Melting Pt: 70 °C Flash Pt: 130 °C |
MDL Number:
MFCD00006936 CAS Number: 105-60-2 EINECS: 203-313-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...