2-Chloro-6-methoxypyridine ≥98.0%

Supplier: TCI America
Warning

C0197-25G C0197-5G
TCC0197-025GEA 69.52 CAD
TCC0197-025G TCC0197-5G
2-Chloro-6-methoxypyridine ≥98.0%
2-Chloro-6-methoxypyridine

CAS Number: 17228-64-7
MDL Number: MFCD00006265
Molecular Formula: C6H6ClNO
Molecular Weight: 143.57
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 184
Flash Point (°C): 76
Specific Gravity (20/20): 1.23

Formula: C₆H₆ClNO
MW: 143.57 g/mol
Boiling Pt: 184 °C
Density: 1.23 (20 °C)
Flash Pt: 76 °C
MDL Number: MFCD00006265
CAS Number: 17228-64-7
EINECS: 241-264-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d443d702897288f81574adbcb25c385a02ec9db&vid=8d443d702897288f81574adbcb25c385a02ec9db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR