2-Chlorolepidine ≥98.0%

Supplier: TCI America
Warning

C0191-5G C0191-25G
TCC0191-5GEA 71.1 CAD
TCC0191-5G TCC0191-025G
2-Chlorolepidine ≥98.0%
2-Chlorolepidine

CAS Number: 634-47-9
MDL Number: MFCD00006742
Molecular Formula: C10H8ClN
Molecular Weight: 177.63
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 296
Melting point (°C): 58


Other Product Names / Synonyms: 2-Chloro-4-methylquinoline

Formula: C₁₀H₈ClN
Boiling Pt: 296 °C
Melting Pt: 58 °C
Density: 1.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006742
CAS Number: 634-47-9
EINECS: 211-209-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db65ae79d7e76ff3653f5e4618aeed014f1e83e8&vid=47d0bc400aaba8d0b08716a59f63e7e5caa941ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR