1-Chloro-3-iodobenzene ≥97.0% stabilized
Supplier: TCI America
Stabilization:
Stabilised with Stabilised
C0190-25G
C0190-5G
TCC0190-025GEA
124.82
CAD
TCC0190-025G
76219-654
1-Chloro-3-iodobenzene ≥97.0% stabilized
1-Chloro-3-iodobenzene
(stabilized with Copper chip)
CAS Number: 625-99-0
MDL Number: MFCD00001046
Molecular Formula: C6H4ClI
Molecular Weight: 238.45
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 228
Flash Point (°C): 101
Specific Gravity (20/20): 1.92
Other Product Names / Synonyms: 3-Chloroiodobenzene
Formula:
C₆H₄ClI Boiling Pt: 228 °C Melting Pt: 54.5 °C Density: 1.92 (20 °C) Flash Pt: 101 °C |
MDL Number:
MFCD00001046 CAS Number: 625-99-0 EINECS: 210-920-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...