Chlorohydroquinone ≥85.0%
Supplier: TCI America
Synonyms:
2-Chlorobenzene-1,4-diol, 2-Chlorohydroquinone
C0185-25G
C0185-500G
TCC0185-025GEA
84.37
CAD
TCC0185-025G
TCC0185-500G
Chlorohydroquinone ≥85.0%
Chlorohydroquinone
CAS Number: 615-67-8
MDL Number: MFCD00002341
Molecular Formula: C6H5ClO2
Molecular Weight: 144.55
Purity/Analysis Method: >85.0% (GC)
Form: Crystal
Boiling point (°C): 263
Melting point (°C): 105
Other Product Names / Synonyms: 1-Chloro-2,5-dihydroxybenzene
Formula:
C₆H₅ClO₂ MW: 144.56 g/mol Boiling Pt: 263 °C Melting Pt: 105 °C |
MDL Number:
MFCD00002341 CAS Number: 615-67-8 EINECS: 210-442-8 UN: 2811 ADR: 6.1 III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...