2-(2-Chloroethoxy)ethanol ≥98.0%

Supplier: TCI America
Warning

C0169-500ML C0169-25ML C0169-100ML
TCC0169-500MLEA 339.7 CAD
TCC0169-500ML TCC0169-025ML TCC0169-100ML
2-(2-Chloroethoxy)ethanol ≥98.0%
2-(2-Chloroethoxy)ethanol

CAS Number: 628-89-7
MDL Number: MFCD00002870
Molecular Formula: C4H9ClO2
Molecular Weight: 124.56
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 185
Flash Point (°C): 107
Specific Gravity (20/20): 1.18
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-(2-Chloroethoxy)ethanol

Formula: C₄H₉ClO₂
MW: 124.57 g/mol
Boiling Pt: 185 °C
Density: 1.18 (20 °C)
Flash Pt: 107 °C
MDL Number: MFCD00002870
CAS Number: 628-89-7
EINECS: 211-059-9
UN: 3334
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4aef7e5d0490dbe29eeac385809f2053ef4b2a37&vid=23112b62472f8a40b58d45e8bc700a15b2b79e44&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR