4-Chlorobenzophenone ≥98.0%
Supplier: TCI America
C0136-25G
C0136-500G
TCC0136-025GEA
30.02
CAD
TCC0136-025G
TCC0136-500G
4-Chlorobenzophenone ≥98.0%
4-Chlorobenzophenone
CAS Number: 134-85-0
MDL Number: MFCD00000622
Molecular Formula: C13H9ClO
Molecular Weight: 216.66
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 170
Melting point (°C): 76
Flash Point (°C): 143
Formula:
C₁₃H₉ClO Boiling Pt: 170 °C Melting Pt: 76 °C Flash Pt: 143 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000622 CAS Number: 134-85-0 EINECS: 205-160-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...