2-Chlorobenzoic acid ≥98.0%
Supplier: TCI America
|
Warning
|
C0133-25G
C0133-500G
TCC0133-025GEA
43.42
CAD
TCC0133-025G
TCC0133-500G
2-Chlorobenzoic acid ≥98.0%
2-Chlorobenzoic acid
CAS Number: 118-91-2
MDL Number: MFCD00002412
Molecular Formula: C7H5ClO2
Molecular Weight: 156.57
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystalline Powder
Boiling point (°C): 285
Melting point (°C): 140
Flash Point (°C): 173
Formula:
C₇H₅ClO₂ Boiling Pt: 285 °C Melting Pt: 140 °C Density: 1.544 g/cm³ (20 °C) Flash Pt: 173 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002412 CAS Number: 118-91-2 EINECS: 204-285-4 Merck Index: 13,02143 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...