3-Chlorobenzoic acid ≥99.0%
Supplier: TCI America
|
Warning
|
C0132-25G
C0132-500G
TCC0132-025GEA
48.98
CAD
TCC0132-025G
TCC0132-500G
3-Chlorobenzoic acid ≥99.0%
3-Chlorobenzoic acid
CAS Number: 535-80-8
MDL Number: MFCD00002491
Molecular Formula: C7H5ClO2
Molecular Weight: 156.57
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 157
Formula:
C₇H₅ClO₂ Boiling Pt: 281 °C (1013 hPa) Melting Pt: 157 °C Density: 1.496 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002491 CAS Number: 535-80-8 EINECS: 208-618-4 Merck Index: 13,02142 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...