4-Chlorobenzaldehyde ≥97.0%

Supplier: TCI America
Warning

C0125-25G C0125-500G C0125-100G
TCC0125-025GEA 54.51 CAD
TCC0125-025G TCC0125-500G TCC0125-100G
4-Chlorobenzaldehyde ≥97.0%
4-Chlorobenzaldehyde

CAS Number: 104-88-1
MDL Number: MFCD00003379
Molecular Formula: C7H5ClO
Molecular Weight: 140.57
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 214
Melting point (°C): 48
Flash Point (°C): 88

Formula: C₇H₅ClO
MW: 140.57 g/mol
Boiling Pt: 214 °C
Melting Pt: 48 °C
Density: 1.196 g/cm³ (20 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00003379
CAS Number: 104-88-1
EINECS: 203-247-4
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef0b25c7330957aa2927d1c5510a987bf6627d37&vid=0960b5168f46f51a8c248f9eb04a0baac89a1560&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR