3-Chlorobenzaldehyde ≥98.0%

Supplier: TCI America
Warning

C0124-25G C0124-500G C0124-100G
TCC0124-025GEA 42.66 CAD
TCC0124-025G TCC0124-500G TCC0124-100G
3-Chlorobenzaldehyde ≥98.0%
3-Chlorobenzaldehyde

CAS Number: 587-04-2
MDL Number: MFCD00003350
Molecular Formula: C7H5ClO
Molecular Weight: 140.57
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 214
Flash Point (°C): 88
Freezing point (°C): 11
Specific Gravity (20/20): 1.24

Formula: C₇H₅ClO
MW: 140.57 g/mol
Boiling Pt: 214 °C
Melting Pt: 17…18 °C
Density: 1.24 (20 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00003350
CAS Number: 587-04-2
EINECS: 209-596-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05f5be99a15f4f563e72257b0f838cc1f3a68a05&vid=125fbb7bf2db3e2e9af6ae0cd24919d4e2f1044b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR