4-Chloroanisole ≥98.0%
Supplier: TCI America
C0122-25G
C0122-500G
TCC0122-025GEA
70.74
CAD
TCC0122-025G
TCC0122-500G
4-Chloroanisole ≥98.0%
4-Chloroanisole
CAS Number: 623-12-1
MDL Number: MFCD00000616
Molecular Formula: C7H7ClO
Molecular Weight: 142.58
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 198
Melting point (°C): -18
Flash Point (°C): 78
Specific Gravity (20/20): 1.18
Formula:
C₇H₇ClO Boiling Pt: 198 °C Melting Pt: –18 °C Density: 1.18 (20 °C) Flash Pt: 78 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000616 CAS Number: 623-12-1 EINECS: 210-772-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...