3,4-Dihydroxybenzoic acid ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: Protocatechuic acid

C0055-25G C0055-100G
TCC0055-025GEA 102.54 CAD
TCC0055-025G TCC0055-100G
3,4-Dihydroxybenzoic acid ≥98.0% (by HPLC)
3,4-Dihydroxybenzoic acid

CAS Number: 99-50-3
MDL Number: MFCD00002509
Molecular Formula: C7H6O4
Molecular Weight: 154.12
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 199


Other Product Names / Synonyms: Catechol-4-carboxylic Acid, Protocatechuic Acid, Pyrocatechol-4-carboxylic Acid

Formula: C₇H₆O₄
MW: 154.12 g/mol
Boiling Pt: 125…135 °C (0.001 mmHg)
Melting Pt: 199 °C
Density: 1.5415 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002509
CAS Number: 99-50-3
EINECS: 202-760-0
Merck Index: 12,08080
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f5bdf8a7ddedc3f7309a8f639eacbf5d65f3a8f&vid=39e221e5847c6207f910b16ac0c13b78f9e0bff1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR