2,3-Dihydroxybenzoic acid ≥98.0%

Supplier: TCI America
Warning

Synonyms: Pyrocatechuic acid

C0054-25G C0054-5G
TCC0054-025GEA 219.62 CAD
TCC0054-025G TCC0054-005G
2,3-Dihydroxybenzoic acid ≥98.0%
2,3-Dihydroxybenzoic acid

CAS Number: 303-38-8
MDL Number: MFCD00002446
Molecular Formula: C7H6O4
Molecular Weight: 154.12
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 205


Other Product Names / Synonyms: Catechol-3-carboxylic Acid, Pyrocatechol-3-carboxylic Acid, o-Pyrocatechuic Acid

Formula: C₇H₆O₄
MW: 154.12 g/mol
Boiling Pt: 339
Melting Pt: 205 °C
Storage Temperature: Ambient
MDL Number: MFCD00002446
CAS Number: 303-38-8
EINECS: 206-139-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3eb19bc0bbed1ee5b2da78a002770a445926e208&vid=b590f884c26f63ab707844051a18de8e971d2566&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR