Trimellitic anhydride ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 1,2,4-Benzenetricarboxylic anhydride, 1,2,4-Benzenetricarboxylic 1,2-anhydride

C0046-25G C0046-500G
TCC0046-025GEA 27.87 CAD
TCC0046-025G TCC0046-500G
Trimellitic anhydride ≥97.0% (by HPLC, titration analysis)
Trimellitic anhydride

CAS Number: 552-30-7
MDL Number: MFCD00005925
Molecular Formula: C9H4O5
Molecular Weight: 192.13
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 245
Melting point (°C): 167
Flash Point (°C): 227


Product Categories: Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 1,2,4-Benzenetricarboxylic 1,2-Anhydride, 4-Carboxyphthalic Anhydride

Formula: C₉H₄O₅
MW: 192.13 g/mol
Boiling Pt: 245 °C
Melting Pt: 167 °C
Flash Pt: 227 °C
MDL Number: MFCD00005925
CAS Number: 552-30-7
EINECS: 209-008-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e360ba82b3c62c0577e43ca2e02b6f94530a82cb&vid=e360ba82b3c62c0577e43ca2e02b6f94530a82cb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR