Ammonium carbamate 97,0

Supplier: TCI America
Warning

Synonyms: Carbamic acid ammonium salt

C0027-500G C0027-25G
TCC0027-500GEA 223.1 CAD
TCC0027-500G TCC0027-025G
Ammonium carbamate 97,0
Ammonium carbamate

CAS Number: 1111-78-0
MDL Number: MFCD00013010
Molecular Formula: CH3NO2
Molecular Weight: 78.07
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 59
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Carbamic Acid Ammonium Salt

Formula: H₂NCO₂NH₄
MW: 78.07 g/mol
Melting Pt: 59 °C
MDL Number: MFCD00013010
CAS Number: 1111-78-0
EINECS: 214-185-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5918c9a89a283b768f62c369c594a222a331665f&vid=5918c9a89a283b768f62c369c594a222a331665f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR