2-Chloro-5-nitropyridine ≥99.0%

Supplier: TCI America
Warning

C0025-25G C0025-5G
TCC0025-025GEA 79 CAD
TCC0025-025G TCC0025-5G
2-Chloro-5-nitropyridine ≥99.0%
2-Chloro-5-nitropyridine

CAS Number: 4548-45-2
MDL Number: MFCD00006240
Molecular Formula: C5H3ClN2O2
Molecular Weight: 158.54
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 110
Flash Point (°C): 158

Formula: C₅H₃ClN₂O₂
Melting Pt: 110 °C
Flash Pt: 158 °C
Storage Temperature: Ambient
MDL Number: MFCD00006240
CAS Number: 4548-45-2
EINECS: 224-908-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77688160bcf8cdba3f488f7bc9515a7e01e7e996&vid=77688160bcf8cdba3f488f7bc9515a7e01e7e996&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR