4-Bromo-2,6-difluorobenzaldehyde ≥98.0%
Supplier: TCI America
B3446-100MG
B3446-1G
TCB3446-100MGEA
26.86
CAD
TCB3446-100MG
TCB3446-1G
4-Bromo-2,6-difluorobenzaldehyde ≥98.0%
4-Bromo-2,6-difluorobenzaldehyde
CAS Number: 537013-51-7
MDL Number: MFCD03094459
Molecular Formula: C7H3BrF2O
Molecular Weight: 221.00
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 77
Formula:
C₇H₃BrF₂O Melting Pt: 77 °C |
MDL Number:
MFCD03094459 CAS Number: 537013-51-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...