X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) ≥98.0% (by HPLC, total nitrogen) for biochemical research

Supplier: TCI America

Synonyms: BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

B3201-1G B3201-200MG
TCB3201-1GEA 347.6 CAD
TCB3201-1G TCB3201-200MG
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) ≥98.0% (by HPLC, total nitrogen) for biochemical research
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)

[for Biochemical Research]
CAS Number: 7240-90-6
MDL Number: MFCD00005666
Molecular Formula: C14H15BrClNO6
Molecular Weight: 408.63
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Specific rotation [a]20/D: -66 deg (C=1, DMF)
Storage Temperature: <0°C


Product Categories: Bioscience, Galactose, ƒÀ-Galactosidase Substrates
Other Product Names / Synonyms: 5-Bromo-4-chloro-3-(beta-D-galactopyranosyloxy)indole, X-Gal

Formula: C₁₄H₁₅BrClNO₆
MW: 408.63 g/mol
Melting Pt: 237…239 °C
Storage Temperature: Freezer
MDL Number: MFCD00005666
CAS Number: 7240-90-6
EINECS: 230-640-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e1806f115e9c07e1525fd20382162b45676c18e&vid=451d1910e04b462eaea9f399e990b57fa3e679d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR