Methyl-4-bromobutyrate ≥98.0%

Supplier: TCI America

B2308-250G B2308-25G
TCB2308-250GEA 696.78 CAD
TCB2308-250G TCB2308-25G
Methyl-4-bromobutyrate ≥98.0%
Methyl 4-bromobutyrate

CAS Number: 4897-84-1
MDL Number: MFCD00041482
Molecular Formula: C5H9BrO2
Molecular Weight: 181.03
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.44


Other Product Names / Synonyms: 4-Bromobutyric Acid Methyl Ester

Formula: C₅H₉BrO₂
Density: 1.44 (20 °C)
MDL Number: MFCD00041482
CAS Number: 4897-84-1
EINECS: 225-523-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5875f00eec1563be2be0428e45714fc9ab74fb74&vid=5875f00eec1563be2be0428e45714fc9ab74fb74&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR