3-Bromo-4-fluorobenzoic acid ≥96.0% (by titrimetric analysis)

Supplier: TCI America
Warning

B2133-5G B2133-1G
TCB2133-5GEA 128.93 CAD
TCB2133-5G TCB2133-001G
3-Bromo-4-fluorobenzoic acid ≥96.0% (by titrimetric analysis)
3-Bromo-4-fluorobenzoic acid

CAS Number: 1007-16-5
MDL Number: MFCD00042463
Molecular Formula: C7H4BrFO2
Molecular Weight: 219.01
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Melting point (°C): 139

Formula: C₇H₄BrFO₂
Melting Pt: 139 °C
MDL Number: MFCD00042463
CAS Number: 1007-16-5
EINECS: 213-751-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7a3beee6e21727b385df268fe7e072b4a0b5f4f&vid=dcc8efaf73e1fff6674615275128a6af680cfe0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR