1-Benzyloxy-4-bromobenzene ≥98.0%
Supplier: TCI America
B2127-5G
B2127-25G
TCB2127-5GEA
39.5
CAD
TCB2127-5G
TCB2127-025G
1-Benzyloxy-4-bromobenzene ≥98.0%
1-Benzyloxy-4-bromobenzene
CAS Number: 6793-92-6
MDL Number: MFCD00028016
Molecular Formula: C13H11BrO
Molecular Weight: 263.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 62
Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: Benzyl 4-Bromophenyl Ether, 4-Benzyloxybromobenzene
Formula:
C₁₃H₁₁BrO Melting Pt: 62 °C |
MDL Number:
MFCD00028016 CAS Number: 6793-92-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...