1-Bromo-4-octylbenzene ≥98.0%

Supplier: TCI America

B2117-5G B2117-25G
TCB2117-005GEA 147.1 CAD
TCB2117-005G TCB2117-25G
1-Bromo-4-octylbenzene ≥98.0%
1-Bromo-4-octylbenzene

CAS Number: 51554-93-9
MDL Number: MFCD00038348
Molecular Formula: C14H21Br
Molecular Weight: 269.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 125
Specific Gravity (20/20): 1.13


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 4-n-Octylbromobenzene

Formula: C₁₄H₂₁Br
Boiling Pt: 125 °C
Density: 1.13 (20 °C)
MDL Number: MFCD00038348
CAS Number: 51554-93-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f28bbfe5e932b284cb3b2401e233afa3b93182d&vid=6f28bbfe5e932b284cb3b2401e233afa3b93182d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR