2-Bromo-6-methylpyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
B2114-25G
B2114-5G
TCB2114-25GEA
246.32
CAD
TCB2114-25G
TCB2114-005G
2-Bromo-6-methylpyridine ≥98.0% (by GC, titration analysis)
2-Bromo-6-methylpyridine
CAS Number: 5315-25-3
MDL Number: MFCD00040743
Molecular Formula: C6H6BrN
Molecular Weight: 172.03
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 102
Flash Point (°C): 97
Specific Gravity (20/20): 1.51
Other Product Names / Synonyms: 6-Bromo-2-picoline, 2-Bromo-6-picoline
Formula:
C₆H₆BrN Boiling Pt: 102 °C Density: 1.51 (20 °C) Flash Pt: 97 °C |
MDL Number:
MFCD00040743 CAS Number: 5315-25-3 EINECS: 226-173-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...