2-Bromo-5-methylpyridine ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2-Bromo-5-picoline

B2113-5G B2113-25G
TCB2113-005GEA 75.84 CAD
TCB2113-005G TCB2113-25G
2-Bromo-5-methylpyridine ≥98.0%
2-Bromo-5-methylpyridine

CAS Number: 3510-66-5
MDL Number: MFCD00209553
Molecular Formula: C6H6BrN
Molecular Weight: 172.03
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 44
Flash Point (°C): 103


Other Product Names / Synonyms: 6-Bromo-3-picoline, 2-Bromo-5-picoline

Formula: C₆H₆BrN
MW: 172.02 g/mol
Boiling Pt: 65
Melting Pt: 152.5
Flash Pt: 103 °C
MDL Number: MFCD00209553
CAS Number: 3510-66-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9a085c619c1e21f86c8c22d6c428e75a0e7e817&vid=e9a085c619c1e21f86c8c22d6c428e75a0e7e817&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR