4-Bromo-3-fluoroaniline ≥98.0%
Supplier: TCI America
|
Warning
|
B2102-5G
B2102-1G
TCB2102-5GEA
104.28
CAD
TCB2102-5G
TCB2102-001G
4-Bromo-3-fluoroaniline ≥98.0%
4-Bromo-3-fluoroaniline
CAS Number: 656-65-5
MDL Number: MFCD00672933
Molecular Formula: C6H5BrFN
Molecular Weight: 190.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 72
Flash Point (°C): 104
Formula:
C₆H₅BrFN Boiling Pt: 231 °C (1013 hPa) Melting Pt: 72 °C Flash Pt: 104 °C |
MDL Number:
MFCD00672933 CAS Number: 656-65-5 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...