1-Bromo-4-fluoro-3-nitrobenzene ≥96.0%
Supplier: TCI America
Synonyms:
4-Bromo-1-fluoro-2-nitrobenzene, 5-Bromo-2-fluoronitrobenzene
CAS Number: 364-73-8
MDL Number: MFCD00129165
Molecular Formula: C6H3BrFNO2
Molecular Weight: 220.00
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 147
Melting point (°C): 19
Flash Point (°C): 110
Specific Gravity (20/20): 1.81
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-Bromo-2-fluoronitrobenzene
Formula:
C₆H₃BrFNO₂ Boiling Pt: 147 °C Melting Pt: 19 °C Density: 1.81 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00129165 CAS Number: 364-73-8 UN: 2810 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...