4-Bromo-2,5-difluoroaniline ≥98.0%
Supplier: TCI America
B2072-5G
B2072-1G
TCB2072-5GEA
102.7
CAD
TCB2072-5G
TCB2072-001G
4-Bromo-2,5-difluoroaniline ≥98.0%
4-Bromo-2,5-difluoroaniline
CAS Number: 112279-60-4
MDL Number: MFCD01861125
Molecular Formula: C6H4BrF2N
Molecular Weight: 208.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 76
Formula:
C₆H₄BrF₂N Melting Pt: 76 °C |
MDL Number:
MFCD01861125 CAS Number: 112279-60-4 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...