5-Bromo-2-iodotoluene ≥98.0%
Supplier: TCI America
B2069-5G
B2069-25G
TCB2069-005GEA
45.82
CAD
TCB2069-005G
TCB2069-025G
5-Bromo-2-iodotoluene ≥98.0%
5-Bromo-2-iodotoluene
CAS Number: 116632-39-4
MDL Number: MFCD00060664
Molecular Formula: C7H6BrI
Molecular Weight: 296.93
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 260
Flash Point (°C): 110
Specific Gravity (20/20): 2.10
Formula:
C₇H₆BrI Boiling Pt: 260 °C Density: 2.10 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00060664 CAS Number: 116632-39-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...