4-Bromo-2-chlorotoluene ≥97.0%

Supplier: TCI America
Warning

B2068-5G B2068-25G
TCB2068-005GEA 39.5 CAD
TCB2068-005G TCB2068-025G
4-Bromo-2-chlorotoluene ≥97.0%
4-Bromo-2-chlorotoluene

CAS Number: 89794-02-5
MDL Number: MFCD00060651
Molecular Formula: C7H6BrCl
Molecular Weight: 205.48
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Specific Gravity (20/20): 1.56

Formula: C₇H₆BrCl
Boiling Pt: 110 °C
Density: 1.56 (20 °C)
MDL Number: MFCD00060651
CAS Number: 89794-02-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e5e1b59026f06056ad36f4ed6d8cc5bbb18481c&vid=6e5e1b59026f06056ad36f4ed6d8cc5bbb18481c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR