3-Bromophenetole ≥98.0%

Supplier: TCI America

B2062-5G B2062-25G
TCB2062-005GEA 52.93 CAD
TCB2062-005G TCB2062-025G
3-Bromophenetole ≥98.0%
3-Bromophenetole

CAS Number: 2655-84-7
MDL Number: MFCD00274287
Molecular Formula: C8H9BrO
Molecular Weight: 201.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 231
Flash Point (°C): 83
Specific Gravity (20/20): 1.41


Other Product Names / Synonyms: 1-Bromo-3-ethoxybenzene

Formula: C₈H₉BrO
Boiling Pt: 231 °C
Density: 1.41 (20 °C)
Flash Pt: 83 °C
MDL Number: MFCD00274287
CAS Number: 2655-84-7
EINECS: 220-186-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=205ccb40a97433d4902e12c573554c0a90b3aa36&vid=205ccb40a97433d4902e12c573554c0a90b3aa36&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR