3-Bromo-4-hydroxybenzonitrile ≥95.0%

Supplier: TCI America

B2037-5G B2037-25G
TCB2037-005GEA 112.34 CAD
TCB2037-005G TCB2037-25G
3-Bromo-4-hydroxybenzonitrile ≥95.0%
3-Bromo-4-hydroxybenzonitrile

CAS Number: 2315-86-8
MDL Number: MFCD00143096
Molecular Formula: C7H4BrNO
Molecular Weight: 198.02
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 156


Other Product Names / Synonyms: 2-Bromo-4-cyanophenol

Formula: C₇H₄BrNO
Boiling Pt: 406 °C (1013 hPa)
Melting Pt: 156 °C
MDL Number: MFCD00143096
CAS Number: 2315-86-8
EINECS: 219-022-9
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=273f40387887294f1c84de16645e10690b211820&vid=273f40387887294f1c84de16645e10690b211820&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR