2-Bromo-6-chlorotoluene ≥98.0%

Supplier: TCI America
Warning

Synonyms: 1-Bromo-3-chloro-2-methylbenzene

B2032-25G B2032-5G
TCB2032-25GEA 180.12 CAD
TCB2032-25G TCB2032-005G
2-Bromo-6-chlorotoluene ≥98.0%
2-Bromo-6-chlorotoluene

CAS Number: 62356-27-8
MDL Number: MFCD00061121
Molecular Formula: C7H6BrCl
Molecular Weight: 205.48
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 95
Specific Gravity (20/20): 1.58

Formula: C₇H₆BrCl
Boiling Pt: 120 °C
Density: 1.58 (20 °C)
Flash Pt: 95 °C
MDL Number: MFCD00061121
CAS Number: 62356-27-8
EINECS: 263-520-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b10cb249ad3bd87d21aafab4eefbdf2c5e08bb1b&vid=7f65d7f11f9ef5692b5a9eb2a38c255d081fd255&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR