1,1'-Bis(diphenylphosphino)ferrocene ≥96.0% (by titrimetric analysis)
Supplier: TCI America
B2027-25G
B2027-1G
B2027-5G
TCB2027-25GEA
274.79
CAD
TCB2027-25G
TCB2027-001G
TCB2027-5G
1,1'-Bis(diphenylphosphino)ferrocene ≥96.0% (by titrimetric analysis)
1,1'-Bis(diphenylphosphino)ferrocene
CAS Number: 12150-46-8
MDL Number: MFCD00001422
Molecular Formula: C34H28FeP2
Molecular Weight: 554.39
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Melting point (°C): 182
Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands, Ligands ([2+2+2]Cycloaddition Reaction)
Other Product Names / Synonyms: 1,1'-Ferrocenebis(diphenylphosphine)
Formula:
C₃₄H₂₈FeP₂ Melting Pt: 182 °C |
MDL Number:
MFCD00001422 CAS Number: 12150-46-8 EINECS: 430-420-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...