2-Bromothioanisole ≥96.0%
Supplier: TCI America
|
Warning
|
Synonyms:
2-Bromophenyl methyl sulfide, 1-Bromo-2-(methylthio)benzene, 2-Bromophenyl methyl sulphide, (2-Bromophenyl)(methyl)sulfane, (2-Bromophenyl)(methyl)sulphane
CAS Number: 19614-16-5
MDL Number: MFCD00000066
Molecular Formula: C7H7BrS
Molecular Weight: 203.10
Purity/Analysis Method: >96.0% (GC)
Form: Liquid
Boiling point (°C): 256
Flash Point (°C): 110
Specific Gravity (20/20): 1.54
Other Product Names / Synonyms: 1-Bromo-2-(methylthio)benzene, 2-Bromophenyl Methyl Sulfide
Formula:
C₇H₇BrS MW: 203.1 g/mol Boiling Pt: 256 °C Melting Pt: -24.5 Density: 1.54 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00000066 CAS Number: 19614-16-5 EINECS: 243-183-4 UN: 3334 ADR: 9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...