Palladium(II) acetylacetonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

B2018-5G B2018-1G
TCB2018-5GEA 1472.56 CAD
TCB2018-5G TCB2018-001G
Palladium(II) acetylacetonate ≥98.0% (by titrimetric analysis)
Palladium (II) acetylacetonate

CAS Number: 14024-61-4
MDL Number: MFCD00000025
Molecular Formula: C10H14O4Pd
Molecular Weight: 304.64
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, Cross-coupling Reaction using Transition Metal Catalysts, C-H Bond Activation Reaction
Other Product Names / Synonyms: Acetylacetone Palladium(II) Salt, Palladium(II) Acetylacetonate

Formula: C₁₀H₁₄O₄Pd
Boiling Pt: ∼190 °C (1013 hPa)
Melting Pt: 190 °C
MDL Number: MFCD00000025
CAS Number: 14024-61-4
EINECS: 237-859-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7ccb094adbdfea59d055bdd02edcee2e332949c&vid=a7ccb094adbdfea59d055bdd02edcee2e332949c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR