3'-Bromo-4'-fluoroacetophenone ≥96.0%
Supplier: TCI America
B2014-5G
B2014-25G
TCB2014-005GEA
131.14
CAD
TCB2014-005G
TCB2014-25G
3'-Bromo-4'-fluoroacetophenone ≥96.0%
3'-Bromo-4'-fluoroacetophenone
CAS Number: 1007-15-4
MDL Number: MFCD00042466
Molecular Formula: C8H6BrFO
Molecular Weight: 217.04
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Boiling point (°C): 240
Melting point (°C): 56
Flash Point (°C): 113
Formula:
C₈H₆BrFO Boiling Pt: 240 °C Melting Pt: 56 °C Flash Pt: 113 °C |
MDL Number:
MFCD00042466 CAS Number: 1007-15-4 EINECS: 213-750-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...