1-Bromo-3-(trifluoromethoxy)benzene ≥98.0%
Supplier: TCI America
B2007-25G
B2007-5G
TCB2007-25GEA
99.22
CAD
TCB2007-25G
TCB2007-005G
1-Bromo-3-(trifluoromethoxy)benzene ≥98.0%
1-Bromo-3-(trifluoromethoxy)benzene
CAS Number: 2252-44-0
MDL Number: MFCD00040943
Molecular Formula: C7H4BrF3O
Molecular Weight: 241.01
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 81
Flash Point (°C): 62
Specific Gravity (20/20): 1.64
Formula:
C₇H₄BrF₃O Boiling Pt: 81 °C Density: 1.64 (20 °C) Flash Pt: 62 °C |
MDL Number:
MFCD00040943 CAS Number: 2252-44-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...