4-Bromo-2-fluorobenzaldehyde ≥96.0%

Supplier: TCI America
Warning

B2002-5G B2002-25G
TCB2002-005GEA 97.96 CAD
TCB2002-005G TCB2002-25G
4-Bromo-2-fluorobenzaldehyde ≥96.0%
4-Bromo-2-fluorobenzaldehyde

CAS Number: 57848-46-1
MDL Number: MFCD00143261
Molecular Formula: C7H4BrFO
Molecular Weight: 203.01
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 63

Formula: C₇H₄BrFO
Melting Pt: 63 °C
MDL Number: MFCD00143261
CAS Number: 57848-46-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e702d3bf4c741f46f4e55152619265b6adb664f6&vid=f3b7565e8f19249aadc14e4b3fce831d6aca4406&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR