1-Bromo-6-chlorohexane ≥95.0%
Supplier: TCI America
B1998-25G
B1998-500G
TCB1998-025GEA
30.02
CAD
TCB1998-025G
TCB1998-500G
1-Bromo-6-chlorohexane ≥95.0%
1-Bromo-6-chlorohexane
CAS Number: 6294-17-3
MDL Number: MFCD00001019
Molecular Formula: C6H12BrCl
Molecular Weight: 199.52
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 96
Flash Point (°C): 117
Specific Gravity (20/20): 1.34
Other Product Names / Synonyms: 6-Bromohexyl Chloride, 6-Chlorohexyl Bromide
Formula:
BrH₂C(CH₂)₄CH₂Cl Boiling Pt: 96 °C Density: 1.34 (20 °C) Flash Pt: 117 °C |
MDL Number:
MFCD00001019 CAS Number: 6294-17-3 EINECS: 228-555-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...